Assay Details
| Gene Symbol: TIMM10B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002754951.4 | XP_002754997.1 | 2 - 3 | 204 | 91 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TIMM10B| Gene Symbol: TIMM10B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002754951.4 | XP_002754997.1 | 2 - 3 | 204 | 91 | |
Set Membership: |
|