Assay Details
| Gene Symbol: BMPR1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002756153.4 | XP_002756199.1 | 11 - 12 | 2002 | 82 | |
| XM_017978950.1 | XP_017834439.1 | 11 - 12 | 1784 | 82 | ||
| XM_017978951.1 | XP_017834440.1 | 12 - 13 | 2014 | 82 | ||
| XM_017978952.1 | XP_017834441.1 | 13 - 14 | 2151 | 82 | ||
| XM_017978953.1 | XP_017834442.1 | 12 - 13 | 2009 | 82 | ||
| XM_017978954.1 | XP_017834443.1 | 10 - 11 | 1597 | 82 | ||
More Information
Set Membership: |
|

BMPR1A
Genomic Map