Assay Details
| Gene Symbol: DNER | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002749868.3 | XP_002749914.1 | 5 - 6 | 1152 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DNER| Gene Symbol: DNER | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002749868.3 | XP_002749914.1 | 5 - 6 | 1152 | 68 | |
Set Membership: |
|