Assay Details
| Gene Symbol: PPP1R3F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017968384.1 | XP_017823873.1 | 4 - 5 | 1189 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PPP1R3F| Gene Symbol: PPP1R3F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017968384.1 | XP_017823873.1 | 4 - 5 | 1189 | 64 | |
Set Membership: |
|