Assay Details
| Gene Symbol: MSI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002753076.3 | XP_002753122.1 | 2 - 3 | 146 | 82 | |
| XM_009004759.2 | XP_009003007.1 | 2 - 3 | 125 | 82 | ||
| XM_009004760.2 | XP_009003008.1 | 2 - 3 | 125 | 82 | ||
| XM_009004761.2 | XP_009003009.1 | 2 - 3 | 125 | 82 | ||
| XM_009004763.2 | XP_009003011.1 | 2 - 3 | 125 | 82 | ||
| XM_009004764.2 | XP_009003012.1 | 2 - 3 | 125 | 82 | ||
| XM_009004765.2 | XP_009003013.1 | 2 - 3 | 125 | 82 | ||
More Information
Set Membership: |
|

MSI1
Genomic Map