Assay Details
| Gene Symbol: FBLN5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017977302.1 | XP_017832791.1 | 10 - 11 | 1394 | 66 | |
| XM_017977303.1 | XP_017832792.1 | 10 - 11 | 1585 | 66 | ||
More Information
Set Membership: |
|

FBLN5
Genomic Map