Assay Details
| Gene Symbol: RPAP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009003588.2 | XP_009001836.1 | 8 - 9 | 1133 | 95 | |
| XM_017975950.1 | XP_017831439.1 | 8 - 9 | 1133 | 95 | ||
More Information
Set Membership: |
|

RPAP3
Genomic Map