Assay Details
| Gene Symbol: PDE6D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002749890.4 | XP_002749936.1 | 2 - 3 | 343 | 91 | |
| XM_008999694.2 | XP_008997942.1 | 2 - 3 | 343 | 91 | ||
More Information
Set Membership: |
|

PDE6D
Genomic Map