Assay Details
Gene Symbol: POU6F1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_009003816.1 | XP_009002064.1 | 5 - 6 | 717 | 68 | |
XM_009003817.2 | XP_009002065.1 | 3 - 4 | 524 | 68 | ||
XM_017976063.1 | XP_017831552.1 | 6 - 7 | 1090 | 68 | ||
XM_017976064.1 | XP_017831553.1 | 6 - 7 | 1085 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
