Assay Details
| Gene Symbol: DDX5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002748148.4 | XP_002748194.1 | 5 - 6 | 754 | 78 | |
| XM_008996915.2 | XP_008995163.1 | 6 - 7 | 865 | 78 | ||
| XM_008996916.2 | XP_008995164.1 | 6 - 7 | 727 | 78 | ||
More Information
Set Membership: |
|

DDX5
Genomic Map