Assay Details
| Gene Symbol: CR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017966629.1 | XP_017822118.1 | 14 - 15 | 2444 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CR1| Gene Symbol: CR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017966629.1 | XP_017822118.1 | 14 - 15 | 2444 | 70 | |
Set Membership: |
|