Assay Details
| Gene Symbol: GFRA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002756615.3 | XP_002756661.1 | 7 - 8 | 1226 | 75 | |
| XM_002756616.3 | XP_002756662.1 | 6 - 7 | 1542 | 75 | ||
| XM_009010334.2 | XP_009008582.1 | 7 - 8 | 1557 | 75 | ||
More Information
Set Membership: |
|

GFRA1
Genomic Map