Assay Details
| Gene Symbol: TAB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008989118.2 | XP_008987366.1 | 4 - 5 | 2299 | 87 | |
| XM_017968580.1 | XP_017824069.1 | 5 - 6 | 2161 | 87 | ||
| XM_017968581.1 | XP_017824070.1 | 6 - 7 | 2255 | 87 | ||
| XM_017968582.1 | XP_017824071.1 | 4 - 5 | 2063 | 87 | ||
| XM_017968583.1 | XP_017824072.1 | 3 - 4 | 2455 | 87 | ||
| XM_017968584.1 | XP_017824073.1 | 5 - 6 | 2162 | 87 | ||
More Information
Set Membership: |
|

TAB3
Genomic Map