Assay Details
| Gene Symbol: KCNG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002747675.3 | XP_002747721.1 | 2 - 3 | 1078 | 80 | |
| XM_008996133.2 | XP_008994381.1 | 3 - 4 | 1374 | 80 | ||
| XM_008996135.2 | XP_008994383.1 | 2 - 3 | 1005 | 80 | ||
| XM_017972113.1 | XP_017827602.1 | 3 - 4 | 3307 | 80 | ||
More Information
Set Membership: |
|

KCNG1
Genomic Map