Assay Details
| Gene Symbol: DHCR24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017974783.1 | XP_017830272.1 | 4 - 5 | 1645 | 92 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DHCR24| Gene Symbol: DHCR24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017974783.1 | XP_017830272.1 | 4 - 5 | 1645 | 92 | |
Set Membership: |
|