Assay Details
| Gene Symbol: HAPLN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002744793.2 | XP_002744839.1 | 4 - 5 | 1212 | 123 | |
| XM_017969802.1 | XP_017825291.1 | 3 - 4 | 999 | 123 | ||
More Information
Set Membership: |
|

HAPLN1
Genomic Map