Assay Details
| Gene Symbol: NT5C1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002750642.3 | XP_002750688.1 | 5 - 6 | 870 | 60 | |
| XM_009000952.2 | XP_008999200.1 | 5 - 6 | 1461 | 60 | ||
More Information
Set Membership: |
|

NT5C1A
Genomic Map