Assay Details
| Gene Symbol: ZDHHC14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008995301.2 | XP_008993549.2 | 4 - 5 | 1324 | 59 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZDHHC14| Gene Symbol: ZDHHC14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008995301.2 | XP_008993549.2 | 4 - 5 | 1324 | 59 | |
Set Membership: |
|