Assay Details
| Gene Symbol: NUMB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009006297.2 | XP_009004545.1 | 8 - 9 | 781 | 75 | |
| XM_009006298.1 | XP_009004546.1 | 7 - 8 | 714 | 75 | ||
| XM_009006299.2 | XP_009004547.1 | 8 - 9 | 857 | 75 | ||
| XM_009006300.2 | XP_009004548.1 | 8 - 9 | 781 | 75 | ||
| XM_009006301.2 | XP_009004549.1 | 7 - 8 | 748 | 75 | ||
| XM_009006302.1 | XP_009004550.1 | 6 - 7 | 681 | 75 | ||
| XM_009006303.1 | XP_009004551.1 | 6 - 7 | 681 | 75 | ||
| XM_009006304.2 | XP_009004552.1 | 8 - 9 | 781 | 75 | ||
| XM_009006305.2 | XP_009004553.1 | 7 - 8 | 748 | 75 | ||
| XM_017977200.1 | XP_017832689.1 | 9 - 10 | 807 | 75 | ||
| XM_017977201.1 | XP_017832690.1 | 7 - 8 | 591 | 75 | ||
| XM_017977202.1 | XP_017832691.1 | 7 - 8 | 733 | 75 | ||
More Information
Set Membership: |
|

NUMB
Genomic Map