Assay Details
| Gene Symbol: ETF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002744206.4 | XP_002744252.1 | 5 - 6 | 764 | 74 | |
| XM_017967971.1 | XP_017823460.1 | 5 - 6 | 958 | 74 | ||
| XM_017967973.1 | XP_017823462.1 | 5 - 6 | 803 | 74 | ||
More Information
Set Membership: |
|

ETF1
Genomic Map