Assay Details
| Gene Symbol: CTNNBIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009000375.2 | XP_008998623.1 | 3 - 4 | 444 | 59 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CTNNBIP1| Gene Symbol: CTNNBIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009000375.2 | XP_008998623.1 | 3 - 4 | 444 | 59 | |
Set Membership: |
|