Assay Details
| Gene Symbol: SP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002751505.3 | XP_002751551.1 | 2 - 3 | 179 | 111 | |
| XM_017975264.1 | XP_017830753.1 | 2 - 3 | 174 | 111 | ||
| XM_017975265.1 | XP_017830754.1 | 2 - 3 | 179 | 111 | ||
More Information
Set Membership: |
|

SP4
Genomic Map