Assay Details
| Gene Symbol: NR1I2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008982382.2 | XP_008980630.1 | 2 - 3 | 755 | 100 | |
| XM_008982383.2 | XP_008980631.1 | 2 - 3 | 610 | 100 | ||
More Information
Set Membership: |
|

NR1I2
Genomic Map