Assay Details
| Gene Symbol: AZI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002759675.4 | XP_002759721.1 | 4 - 5 | 618 | 69 | |
| XM_017965910.1 | XP_017821399.1 | 4 - 5 | 759 | 69 | ||
| XM_017965911.1 | XP_017821400.1 | 5 - 6 | 704 | 69 | ||
More Information
Set Membership: |
|

AZI2
Genomic Map