Assay Details
| Gene Symbol: GNA13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002748656.4 | XP_002748702.1 | 3 - 4 | 703 | 119 | |
| XM_017972756.1 | XP_017828245.1 | 4 - 5 | 761 | 119 | ||
More Information
Set Membership: |
|

GNA13
Genomic Map