Assay Details
| Gene Symbol: EXOSC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002743400.3 | XP_002743446.1 | 4 - 5 | 420 | 73 | |
| XM_009004266.2 | XP_009002514.1 | 4 - 5 | 339 | 73 | ||
More Information
Set Membership: |
|

EXOSC2
Genomic Map