Assay Details
| Gene Symbol: MALT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002757258.4 | XP_002757304.2 | 10 - 11 | 2260 | 109 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MALT1| Gene Symbol: MALT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002757258.4 | XP_002757304.2 | 10 - 11 | 2260 | 109 | |
Set Membership: |
|