Assay Details
| Gene Symbol: FST | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002744986.4 | XP_002745032.1 | 2 - 3 | 649 | 99 | |
| XM_002744987.4 | XP_002745033.1 | 2 - 3 | 650 | 99 | ||
| XM_008992077.2 | XP_008990325.1 | 2 - 3 | 650 | 99 | ||
| XM_008992078.2 | XP_008990326.1 | 2 - 3 | 653 | 99 | ||
More Information
Set Membership: |
|

FST
Genomic Map