Assay Details
| Gene Symbol: WDR70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002745045.3 | XP_002745091.3 | 9 - 10 | 1145 | 116 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

WDR70| Gene Symbol: WDR70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002745045.3 | XP_002745091.3 | 9 - 10 | 1145 | 116 | |
Set Membership: |
|