Assay Details
| Gene Symbol: RBFOX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017963417.1 | XP_017818906.1 | 10 - 11 | 1357 | 115 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RBFOX1| Gene Symbol: RBFOX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017963417.1 | XP_017818906.1 | 10 - 11 | 1357 | 115 | |
Set Membership: |
|