Assay Details
| Gene Symbol: YARS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002752379.4 | XP_002752425.1 | 4 - 5 | 1858 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

YARS2| Gene Symbol: YARS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002752379.4 | XP_002752425.1 | 4 - 5 | 1858 | 65 | |
Set Membership: |
|