Assay Details
| Gene Symbol: LEO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002753491.3 | XP_002753537.1 | 9 - 10 | 1676 | 71 | |
| XM_009005455.1 | XP_009003703.1 | 9 - 10 | 1742 | 71 | ||
| XM_009005456.1 | XP_009003704.1 | 8 - 9 | 1607 | 71 | ||
More Information
Set Membership: |
|

LEO1
Genomic Map