Assay Details
| Gene Symbol: NRG3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009009628.2 | XP_009007876.2 | 5 - 6 | 1174 | 69 | |
| XM_009009629.2 | XP_009007877.2 | 5 - 6 | 1177 | 69 | ||
| XM_017963418.1 | XP_017818907.1 | 5 - 6 | 1177 | 69 | ||
| XM_017963419.1 | XP_017818908.1 | 5 - 6 | 1177 | 69 | ||
| XM_017963420.1 | XP_017818909.1 | 5 - 6 | 1174 | 69 | ||
| XM_017963421.1 | XP_017818910.1 | 5 - 6 | 1177 | 69 | ||
More Information
Set Membership: |
|

NRG3
Genomic Map