Assay Details
| Gene Symbol: KCNN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002760046.4 | XP_002760092.1 | 4 - 5 | 1837 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

KCNN3| Gene Symbol: KCNN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002760046.4 | XP_002760092.1 | 4 - 5 | 1837 | 72 | |
Set Membership: |
|