Assay Details
| Gene Symbol: WSB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009004749.2 | XP_009002997.1 | 4 - 5 | 717 | 60 | |
| XM_017976407.1 | XP_017831896.1 | 3 - 4 | 583 | 60 | ||
More Information
Set Membership: |
|

WSB2
Genomic Map