Assay Details
| Gene Symbol: ERG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002761421.3 | XP_002761467.1 | 10 - 11 | 1265 | 81 | |
| XM_002761422.3 | XP_002761468.1 | 8 - 9 | 1119 | 81 | ||
| XM_008986692.2 | XP_008984940.1 | 9 - 10 | 1194 | 81 | ||
| XM_017967151.1 | XP_017822640.1 | 8 - 9 | 1013 | 81 | ||
More Information
Set Membership: |
|

ERG
Genomic Map