Assay Details
| Gene Symbol: KANSL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002752402.4 | XP_002752448.2 | 2 - 3 | 720 | 64 | |
| XM_009003701.2 | XP_009001949.1 | 2 - 3 | 720 | 64 | ||
More Information
Set Membership: |
|

KANSL2
Genomic Map