Assay Details
| Gene Symbol: SLC7A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008989458.2 | XP_008987706.1 | 2 - 3 | 558 | 84 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC7A3| Gene Symbol: SLC7A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008989458.2 | XP_008987706.1 | 2 - 3 | 558 | 84 | |
Set Membership: |
|