Assay Details
| Gene Symbol: NECTIN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003735162.3 | XP_003735210.1 | 7 - 8 | 2317 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NECTIN4| Gene Symbol: NECTIN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003735162.3 | XP_003735210.1 | 7 - 8 | 2317 | 71 | |
Set Membership: |
|