Assay Details
| Gene Symbol: AUTS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008981354.2 | XP_008979602.1 | 1 - 2 | 1435 | 69 | |
| XM_017970021.1 | XP_017825510.1 | 1 - 2 | 1435 | 69 | ||
More Information
Set Membership: |
|

AUTS2
Genomic Map