Assay Details
| Gene Symbol: TRIB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002758023.3 | XP_002758069.1 | 1 - 2 | 1691 | 93 | |
| XM_008981118.2 | XP_008979366.1 | 2 - 3 | 613 | 93 | ||
| XM_017964533.1 | XP_017820022.1 | 2 - 3 | 333 | 93 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008981118.2, XM_017964533.1 transcripts.
Set Membership: |
|

TRIB2
Genomic Map