Assay Details
| Gene Symbol: SH2D1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002763236.3 | XP_002763282.1 | 2 - 3 | 505 | 100 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SH2D1A| Gene Symbol: SH2D1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002763236.3 | XP_002763282.1 | 2 - 3 | 505 | 100 | |
Set Membership: |
|