Assay Details
| Gene Symbol: LOC100412777 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002758109.4 | XP_002758155.1 | 3 - 4 | 522 | 93 | |
| XM_008981310.2 | XP_008979558.1 | 3 - 4 | 522 | 93 | ||
| XM_017964713.1 | XP_017820202.1 | 3 - 4 | 522 | 93 | ||
| XM_017964714.1 | XP_017820203.1 | 3 - 4 | 521 | 93 | ||
| XM_017964715.1 | XP_017820204.1 | 3 - 4 | 522 | 93 | ||
| XM_017964716.1 | XP_017820205.1 | 3 - 4 | 522 | 93 | ||
More Information
Set Membership: |
|

LOC100412777
Genomic Map