Assay Details
| Gene Symbol: DMWD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017967665.1 | XP_017823154.1 | 4 - 5 | 1909 | 88 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

DMWD| Gene Symbol: DMWD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017967665.1 | XP_017823154.1 | 4 - 5 | 1909 | 88 | |