Assay Details
| Gene Symbol: INSYN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002756710.3 | XP_002756756.1 | 3 - 4 | 1411 | 77 | |
| XM_017963272.1 | XP_017818761.1 | 2 - 3 | 2839 | 77 | ||
More Information
Set Membership: |
|

INSYN2
Genomic Map