Assay Details
| Gene Symbol: SLC6A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008997142.1 | XP_008995390.1 | 12 - 13 | 2114 | 59 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC6A4| Gene Symbol: SLC6A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008997142.1 | XP_008995390.1 | 12 - 13 | 2114 | 59 | |
Set Membership: |
|