Assay Details
| Gene Symbol: NDFIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002744294.3 | XP_002744340.1 | 5 - 6 | 927 | 99 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NDFIP1| Gene Symbol: NDFIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002744294.3 | XP_002744340.1 | 5 - 6 | 927 | 99 | |
Set Membership: |
|