Assay Details
| Gene Symbol: RANBP3L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002745050.3 | XP_002745096.1 | 12 - 13 | 1777 | 148 | |
| XM_017969956.1 | XP_017825445.1 | 12 - 13 | 1779 | 148 | ||
| XM_017969957.1 | XP_017825446.1 | 13 - 14 | 1854 | 148 | ||
More Information
Set Membership: |
|

RANBP3L
Genomic Map