Assay Details
| Gene Symbol: NABP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002749575.4 | XP_002749621.1 | 3 - 4 | 792 | 104 | |
| XM_008999122.2 | XP_008997370.1 | 3 - 4 | 392 | 104 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008999122.2 transcripts.
Set Membership: |
|

NABP1
Genomic Map