Assay Details
| Gene Symbol: PRDX4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002762725.3 | XP_002762771.1 | 1 - 2 | 378 | 69 | |
| XM_008989076.2 | XP_008987324.1 | 1 - 2 | 378 | 69 | ||
| XM_008989077.2 | XP_008987325.1 | 1 - 2 | 378 | 69 | ||
More Information
Set Membership: |
|

PRDX4
Genomic Map